Anti-IgG (H+L) Goat Polyclonal Antibody (Cy2®)

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK710-111-032EA 1750 DKK
ROCK710-111-032
Anti-IgG (H+L) Goat Polyclonal Antibody (Cy2®)
Antistoffer
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Cy2®
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR