Anti-IgA Goat Polyclonal Antibody (Biotin)

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK709-1606EA 3720 DKK
ROCK709-1606
Anti-IgA Goat Polyclonal Antibody (Biotin)
Antistoffer
This product is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms.

Recommended Dilutions: ELISA: 1:20000 - 1:400,000; Flow Cytometry: 1:200 - 1:1000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:500 - 1:5000; 1:200 - 1:1000 for flow cytometry and fluorescence immunochemistry
Restore with deionized water (or equivalent); 500 µl

Type: Secondary
Antigen: IgA α
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR