Anti-kappa light chain Goat Polyclonal Antibody (Peroxidase)
Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ROCK709-1310EA 3720 DKK
ROCK709-1310
Anti-kappa light chain Goat Polyclonal Antibody (Peroxidase)
Antistoffer
F(ab')2 Anti-human kappa chain has been assayed against 1.0 µg of human kappa light chain in a standard capture ELISA using ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid)) as a substrate for 30 minutes at room temperature.
Recommended Dilutions: ELISA: 1:50000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: Igκ
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Light Chain
Host: Goat
Isotype:
Reactivity: Human