Anti-IgM Fc5µ Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK709-1231EA 1320 DKK
ROCK709-1231
Anti-IgM Fc5µ Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: Immunofluorescence: 1:500 - 1:2500; Flow Cytometry: 1:500-1:2500;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgM Fc5µ
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: Fc5µ
Host: Goat
Isotype: IgM
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR