Anti-IgG (H+L) Goat Polyclonal Antibody
Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ROCK709-1116EA 2070 DKK
ROCK709-1116
Anti-IgG (H+L) Goat Polyclonal Antibody
Antistoffer
F(ab')2 Anti-Human IgG (H&L) Antibody is suitable for immunoblotting (western or dot blot), ELISA, and immunochemistry as well as other peroxidase-antibody based enzymatic assays requiring lot-to-lot consistency.
Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Human