Anti-Biotin Goat Polyclonal Antibody (Cy2®)
Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ROCK700-111-098EA 7280 DKK
ROCK700-111-098
Anti-Biotin Goat Polyclonal Antibody (Cy2®)
Antistoffer
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.
Recommended Dilutions: Immunofluorescence: 1:1000-1:5000; Flow Cytometry: 1:500-1:2500
Restore with deionized water (or equivalent); 1.0 ml
Type: Tag
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: Cy2®
Epitope:
Host: Goat
Isotype: IgG
Reactivity: pan