Anti-Biotin Goat Polyclonal Antibody (Cy3®)

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK700-104-098EA 3320 DKK
ROCK700-104-098
Anti-Biotin Goat Polyclonal Antibody (Cy3®)
Antistoffer
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.

Recommended Dilutions: Immunofluorescence: 1:100-1:1000; Flow Cytometry: 1:200-1:400; Flow Cytometry: 1:200-1:400
Restore with deionized water (or equivalent); 1.0 ml

Type: Tag
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Goat
Isotype: IgG
Reactivity: pan
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR