Anti-IgM µ Goat Polyclonal Antibody (Texas Red®)

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK618-109-007EA 2370 DKK
ROCK618-109-007
Anti-IgM µ Goat Polyclonal Antibody (Texas Red®)
Antistoffer
Secondary Goat Anti-IgM (mu chain) Reacts with Ferret

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: µ-chain
Host: Goat
Isotype: IgM
Reactivity: Ferret
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR