Anti-IgM µ Goat Polyclonal Antibody (Biotin)

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK618-106-007EA 1160 DKK
ROCK618-106-007
Anti-IgM µ Goat Polyclonal Antibody (Biotin)
Antistoffer
Secondary Goat Anti-IgM (mu chain) Reacts with Ferret

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: µ-chain
Host: Goat
Isotype: IgM
Reactivity: Ferret
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR