Anti-IgA Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK618-102-006EA 1080 DKK
ROCK618-102-006
Anti-IgA Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Secondary Goat Anti-IgA Reacts with Ferret

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgA α
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Goat
Isotype: IgA
Reactivity: Ferret
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR