Anti-IgA/IgG/IgM Goat Polyclonal Antibody (Biotin)

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK617-106-130EA 1360 DKK
ROCK617-106-130
Anti-IgA/IgG/IgM Goat Polyclonal Antibody (Biotin)
Antistoffer
Secondary Goat Anti-IgG IgA IgM Reacts with Monkey

Recommended Dilutions: ELISA: 1:50000 - 1:100000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:200 - 1:1000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgA/IgG/IgM
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgA
Reactivity: Monkey
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR