Anti-IgA Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK617-105-006EA 1750 DKK
ROCK617-105-006
Anti-IgA Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Secondary Goat Anti-IgA Reacts with Monkey

Recommended Dilutions: ELISA: 1:2000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgA α
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgA
Reactivity: Monkey
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR