Anti-IgG (H+L) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK616-1502EA 2480 DKK
ROCK616-1502
Anti-IgG (H+L) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antistoffer
Suitable for immunoblotting (western or dot blot), ELISA and immunochemistry as well as other phosphatase-antibody based enzymatic assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Donkey
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR