Anti-IgM µ Rabbit Polyclonal Antibody (Biotin)

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK612-4607EA 1440 DKK
ROCK612-4607
Anti-IgM µ Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Anti-Rat IgM (mu chain) Antibody has been assayed against Rat IgM in a standard capture ELISA using Peroxidase Conjugated Streptavidin.  A working dilution of 1:15,000 to 1:70,000 of the  concentration is suggested for this product.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: µ-chain
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR