Anti-IgG (H+L) Rabbit Polyclonal Antibody

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK612-4126EA 1070 DKK
ROCK612-4126
Anti-IgG (H+L) Rabbit Polyclonal Antibody
Antistoffer
Antibody anti-Rat has been assayed against 1.0 ug of Rat IgG in a standard capture ELISA using ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid]) code # ABTS-100 as a substrate for 30 minutes at room temperature. A working dilution of 1:2,000 to

Recommended Dilutions: ELISA: 1:20000 - 1:300000; Western Blot: 1:2000-1:10000; Immunochemistry: 1:1000-1:5000; Other Dilution: User Optimized

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR