Anti-IgG F(ab')2 Rabbit Polyclonal Antibody

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK612-4104EA 1730 DKK
ROCK612-4104
Anti-IgG F(ab')2 Rabbit Polyclonal Antibody
Antistoffer
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR