Anti-IgA/IgG/IgM Goat Polyclonal Antibody (Biotin)

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK612-106-130EA 1470 DKK
ROCK612-106-130
Anti-IgA/IgG/IgM Goat Polyclonal Antibody (Biotin)
Antistoffer
Anti-Rat IgG IgA IgM (H&L) Biotin conjugated antibodies are suitable for immunoblotting (western or dot blot), ELISA, immunoperoxidase electron microscopy and immunochemistry as well as other peroxidase-antibody based enzymatic assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:150000; Western Blot: 1:2000 - 1:20000; Immunochemistry: 1:1000 - 1:10000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Secondary
Antigen: IgA/IgG/IgM
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgA
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR