Anti-IgG (H+L) Donkey Polyclonal Antibody
Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ROCK611-701-127EA 2070 DKK
ROCK611-701-127
Anti-IgG (H+L) Donkey Polyclonal Antibody
Antistoffer
Anti-Rabbit IgG (H&l) is suitable for use in immunoelectrophoresis, western-blot, competitive western-blot, ELISA and competitive ELISA assays. Specific conditions for reactivity and signal detection should be optimized by the end user.
Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Rabbit