Anti-IgG (H+L) Goat Polyclonal Antibody

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK611-101-122EA 1060 DKK
ROCK611-101-122
Anti-IgG (H+L) Goat Polyclonal Antibody
Antistoffer
Secondary Goat Anti-IgG (H&L) Reacts with Rabbit (Lapine)

Recommended Dilutions: ELISA: 1:15000 - 1:30,000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:500 - 1:2000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR