Anti-IgG (H+L) Donkey Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK610-705-124EA 2040 DKK
ROCK610-705-124
Anti-IgG (H+L) Donkey Polyclonal Antibody (AP (Alkaline Phosphatase))
Antistoffer
Secondary Donkey Anti-IgG (H&L) Reacts with Mouse

Recommended Dilutions: ELISA: 1:2000 - 1:20000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR