Anti-IgG (H+L) Donkey Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK610-702-002EA 866 DKK
ROCK610-702-002
Anti-IgG (H+L) Donkey Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Secondary Donkey Anti-IgG (H&L) Reacts with Mouse

Recommended Dilutions:
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR