Anti-IgG (Human) (human IgG) Goat Polyclonal Antibody (IRDye® 700DX)

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK609-130-123EA 3310 DKK
ROCK609-130-123
Anti-IgG (Human) (human IgG) Goat Polyclonal Antibody (IRDye® 700DX)
Fluorescence technology is widely used to detect Proteins. However, many common visible fluorophores often result in considerable background fluorescence in the visible range. Visible fluorophores are rarely used for membrane-based Protein detection.

Recommended Dilutions: FLISA: 1:5000 - 1:10000; Western Blot: 1:10000 - 1:25000;
Restore with deionized water (or equivalent); 500 µl

Recommended Dilutions: FLISA: 1:5000 - 1:10000; Western Blot: 1:10000 - 1:25000;
Restore with deionized water (or equivalent); 500 µl

Type: Secondary
Antigen: IgG (Human) (human IgG)
Clonality: Polyclonal
Clone:
Conjugation: IRDye700DX®
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR