Anti-IgA/IgG/IgM Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK609-105-130EA 1570 DKK
ROCK609-105-130
Anti-IgA/IgG/IgM Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antistoffer
Secondary Goat Anti-IgG IgA IgM Reacts with Human

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:3000; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgA/IgG/IgM
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgA
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR