Anti-IgG (H+L) Goat Polyclonal Antibody

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK608-1102EA 5840 DKK
ROCK608-1102
Anti-IgG (H+L) Goat Polyclonal Antibody
Antistoffer
Type: Secondary
Antigen: IgG (Horse) (horse IgG)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Horse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR