Anti-IgG (H+L) Goat Polyclonal Antibody (Cy5®)

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK607-110-130EA 1810 DKK
ROCK607-110-130
Anti-IgG (H+L) Goat Polyclonal Antibody (Cy5®)
Type: Secondary
Antigen: IgG (Golden Syrian Hamster) (Golden Syrian Hamster IgG)
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Goat
Isotype: none
Reactivity: Hamster
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR