Anti-IgG (H+L) Goat Polyclonal Antibody (Cy5.5®)

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK606-113-129EA 1800 DKK
ROCK606-113-129
Anti-IgG (H+L) Goat Polyclonal Antibody (Cy5.5®)
Antistoffer
Type: Secondary
Antigen: IgG (Guinea Pig) (Guinea Pig IgG)
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR