Anti-IgG F(c) Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK605-4503EA 1490 DKK
ROCK605-4503
Anti-IgG F(c) Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antistoffer
Secondary Rabbit Anti-IgG F(c) Reacts with Goat

Recommended Dilutions: ELISA: 1:4000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: F(c)
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR