Anti-IgG (H+L) Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK605-4002EA 1980 DKK
ROCK605-4002
Anti-IgG (H+L) Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antistoffer
Secondary Rabbit Anti-IgG (H&L) Reacts with Goat

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR