Anti-IgG F(c) Rabbit Polyclonal Antibody (Biotin)

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK603-4603EA 1390 DKK
ROCK603-4603
Anti-IgG F(c) Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Anti-Chicken IgG F(c) antibody is suitable for ELISA,Western Blot and Immunohistrochemistry applications. Specific conditions for reactivity should be optimized by end user.

Recommended Dilutions: ELISA: 1:250000; Western Blot: 1:2000 - 1:20000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: F(c)
Host: Rabbit
Isotype: IgG
Reactivity: Chicken
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR