Anti-IgG (H+L) Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK601-1302EA 1120 DKK
ROCK601-1302
Anti-IgG (H+L) Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Anti-Bovine IgG (heavy and light chain) Antibody is peroxidase conjugated and is suitable for immunoassays where specificity to bovine immunoglobulin is desired. Antibody has been tested by ELISA, western blot immunoblot, and immunohistochemisty.

Recommended Dilutions: ELISA: 1:10000 - 1:200,000; Western Blot: 1:5000 - 1:100000; Immunochemistry: 1:500 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Bovine
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR