Anti-COL4A1 Rabbit Polyclonal Antibody (Biotin)

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK600-406-106EA 2940 DKK
ROCK600-406-106
Anti-COL4A1 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Anti-Collagen Type IV is suitable for western blotting, IHC and for ELISA. Researchers should determine optimal titers for applications that are not stated below.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: COL4A1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Bovine, Mammalian
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR