Anti-IRAK4 Rabbit Polyclonal Antibody

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK600-401-953EA 2830 DKK
ROCK600-401-953
Anti-IRAK4 Rabbit Polyclonal Antibody
Antistoffer
This affinity-purified antibody has been tested for use in ELISA, immunochemistry and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 50 kDa in size corresponding to IRAK-4 Protein.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:250 - 1:1000; Immunochemistry: 1:100 - 1:500;

Type: Primary
Antigen: IRAK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR