Anti-IRS1 Rabbit Polyclonal Antibody

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROCK600-401-445EA 5840 DKK
ROCK600-401-445
Anti-IRS1 Rabbit Polyclonal Antibody
Antistoffer
This affinity purified antibody has been tested for use in ELISA and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 130 kDa in size corresponding to phosphorylated IRS1 Protein by western blotting.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:250 - 1:1500;

Type: Primary
Antigen: IRS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR