Anti-CDH11 Goat Polyclonal Antibody

Leverandør: Everest Biotechnology
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

EVBIEB10072EA 2430 DKK
EVBIEB10072
Anti-CDH11 Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CDH11 (cadherin 11, type 2, OB-cadherin (osteoblast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR