Anti-PSMD10 Rabbit Polyclonal Antibody
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PIEAPA116904EA 0 DKK
PIEAPA116904
Anti-PSMD10 Rabbit Polyclonal Antibody
Antistoffer
Suggested positive control: Hela whole cell extract, antigen standard for PSMD10 (transient overexpression lysate). Gankyrin is an ankyrin-repeat liver oncoprotein that plays an important role in controlling the functions of the tumour suppressors RB and p53.
Type: Primary
Antigen: PSMD10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human