Anti-OLLAS Epitope Tag Rat Monoclonal Antibody (DyLight® 680) [clone: L2]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNBP1-06713FREA 4900 DKK
NOVUNBP1-06713FR
Anti-OLLAS Epitope Tag Rat Monoclonal Antibody (DyLight® 680) [clone: L2]
Antistoffer
Type: Tag
Antigen: OLLAS Epitope Tag
Clonality: monoclonal
Clone: L2
Conjugation: DyLight® 680
Epitope:
Host: Rat
Isotype: IgG1 Kappa
Reactivity: Pan
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR