Anti-BIRC5 Mouse Monoclonal Antibody (DyLight® 755) [clone: 8E2]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB500-644IREA 4790 DKK
NOVUNB500-644IR
Anti-BIRC5 Mouse Monoclonal Antibody (DyLight® 755) [clone: 8E2]
Antistoffer
Type: Primary
Antigen: BIRC5 (baculoviral IAP repeat containing 5)
Clonality: monoclonal
Clone: 800
Conjugation: DyLight® 755
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR