Anti-NQO1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: A180]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB200-209HEA 5360 DKK
NOVUNB200-209H
Anti-NQO1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: A180]
Antistoffer
Type: Primary
Antigen: NQO1 (NAD(P)H dehydrogenase, quinone 1)
Clonality: monoclonal
Clone: A180
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR