Anti-CDKN2A Rat Monoclonal Antibody (DyLight® 755) [clone: 5-C3-1]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB200-174IREA 4760 DKK
NOVUNB200-174IR
Anti-CDKN2A Rat Monoclonal Antibody (DyLight® 755) [clone: 5-C3-1]
Antistoffer
Type: Primary
Antigen: CDKN2A (cyclin-dependent kinase inhibitor 2A)
Clonality: monoclonal
Clone: 5-C3-1
Conjugation: DyLight® 755
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR