Anti-CDKN2A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB200-158HEA 5440 DKK
NOVUNB200-158H
Anti-CDKN2A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Type: Primary
Antigen: CDKN2A (cyclin-dependent kinase inhibitor 2A)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR