Anti-MYC Mouse Monoclonal Antibody (DyLight® 550) [clone: 9E11]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB200-108REA 5400 DKK
NOVUNB200-108R
Anti-MYC Mouse Monoclonal Antibody (DyLight® 550) [clone: 9E11]
Antistoffer
Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: monoclonal
Clone: 900000000000
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR