Anti-TP53 Mouse Monoclonal Antibody (DyLight® 405) [clone: PAb 240]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB200-103VEA 4980 DKK
NOVUNB200-103V
Anti-TP53 Mouse Monoclonal Antibody (DyLight® 405) [clone: PAb 240]
Antistoffer
Type: Primary
Antigen: TP53 (tumor protein p53)
Clonality: monoclonal
Clone: PAb 240
Conjugation: DyLight® 405
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR