Anti-MAT2A Rabbit Polyclonal Antibody (DyLight® 405)

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB110-94158VEA 5290 DKK
NOVUNB110-94158V
Anti-MAT2A Rabbit Polyclonal Antibody (DyLight® 405)
Antistoffer
Type: Primary
Antigen: MAT2A (methionine adenosyltransferase II, alpha)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR