Anti-ITGAM Rabbit Polyclonal Antibody (DyLight® 405)

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB110-89474VEA 5900 DKK
NOVUNB110-89474V
Anti-ITGAM Rabbit Polyclonal Antibody (DyLight® 405)
Antistoffer
Type: Primary
Antigen: ITGAM (integrin, alpha M (complement component 3 receptor 3 subunit))
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR