Anti-SPP1 Mouse Monoclonal Antibody (Biotin) [clone: 1B20]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB110-89062BEA 4980 DKK
NOVUNB110-89062B
Anti-SPP1 Mouse Monoclonal Antibody (Biotin) [clone: 1B20]
Antistoffer
Type: Primary
Antigen: SPP1 (secreted phosphoprotein 1)
Clonality: monoclonal
Clone: 1B20
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR