Anti-TRPM2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB110-82364HEA 5290 DKK
NOVUNB110-82364H
Anti-TRPM2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Type: Primary
Antigen: TRPM2 (transient receptor potential cation channel, subfamily M, member 2)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR