Anti-HSD3B1 Mouse Monoclonal Antibody (DyLight® 550) [clone: FDO66Q]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB110-78644REA 5290 DKK
NOVUNB110-78644R
Anti-HSD3B1 Mouse Monoclonal Antibody (DyLight® 550) [clone: FDO66Q]
Antistoffer
Type: Primary
Antigen: HSD3B1 (hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1)
Clonality: monoclonal
Clone: FDO66Q
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR