Anti-LMO2 Mouse Monoclonal Antibody (DyLight® 680) [clone: 1A9-3B11]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB110-78626FREA 5290 DKK
NOVUNB110-78626FR
Anti-LMO2 Mouse Monoclonal Antibody (DyLight® 680) [clone: 1A9-3B11]
Antistoffer
Type: Primary
Antigen: LMO2 (LIM domain only 2 (rhombotin-like 1))
Clonality: monoclonal
Clone: 1A9-3B11
Conjugation: DyLight® 680
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR