Anti-ATG16L1 Rabbit Polyclonal Antibody (DyLight® 488)

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB110-60928GEA 4900 DKK
NOVUNB110-60928G
Anti-ATG16L1 Rabbit Polyclonal Antibody (DyLight® 488)
Antistoffer
Type: Primary
Antigen: ATG16L1 (autophagy related 16-like 1 (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR