Anti-APOE Mouse Monoclonal Antibody (DyLight® 488) [clone: WUE-4]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB110-60531GEA 5900 DKK
NOVUNB110-60531G
Anti-APOE Mouse Monoclonal Antibody (DyLight® 488) [clone: WUE-4]
Antistoffer
Type: Primary
Antigen: APOE (apolipoprotein E)
Clonality: monoclonal
Clone: WUE-4
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR